missing translation for 'onlineSavingsMsg'
Learn More

Pref-1/DLK1/FA1 Antibody (1168B) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30171277 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30171277 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30171277 Supplier Novus Biologicals Supplier No. FAB8634JF525
  • ONLINE EXCLUSIVE PRICE
    5870.00 NOK
    5551.35 NOK / 0.1mL
    Save 318.65 NOK
    5% Off
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pref-1/DLK1/FA1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1168B
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Pref-1/DLK1/FA1, Ala24-Gln305, Accession # Q09163
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8788
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.