Learn More
Zwiebel™ Precision OIML F1 Weight
Product Code. 11700793
Click to view available options
:
Precision OIML F1 Weight
Product Code. 11700793
Brand: Zwiebel™ PBI23130
Only null left
Add to basket
Add to basket
Boxed weight, certified for precision calibration
  • Precision calibration weight
Specifications
Product Type Weight
Classification OIML F1
Weight (Metric) 2 g
For Use With (Application) Balance calibration