missing translation for 'onlineSavingsMsg'
Learn More

Prealbumin/transthyretin Mouse anti-Human, Mouse, Clone: 1H8C4, Proteintech

Código de producto. p-7213457 Tienda Proteintech Productos
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
150 μL
20 μL
missing translation for 'unitSize'
150µL
20µL
Código de producto. Quantity unitSize
16847685 20 μL 20µL
16837685 150 μL 150µL
2 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16847685

missing translation for 'mfr': Proteintech 661081IG20UL

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Transthyretin, one of the three prealbumins including alpha-1-antitrypsin, transthyretin and orosomucoid. Transthyretin is a carrier protein; it transports thyroid hormones in the plasma and cerebrospinal fluid, and also transports retinol (vitamin A) in the plasma. The protein consists of a tetramer of identical subunits. More than 80 different mutations in this gene have been reported; most mutations are related to amyloid deposition, affecting predomitly peripheral nerve and/or the heart, and a small portion of the gene mutations is non-amyloidogenic. The diseases caused by mutations include amyloidotic polyneuropathy, euthyroid hyperthyroxinaemia, amyloidotic vitreous opacities, cardiomyopathy, oculoleptomeningeal amyloidosis, meningocerebrovascular amyloidosis, carpal tunnel syndrome, etc.

Especificaciones

Antigen Prealbumin/transthyretin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1H8C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TTR
Gene Accession No. P02766, P07309
Gene Alias ATTR, HsT2651, PALB, Prealbumin, Prealbumin/transthyretin, TBPA, transthyretin, TTR
Gene Symbols TTR
Host Species Mouse
Immunogen Prealbumin/transthyretin Fusion Protein Ag19232
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22139, 7276
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.