Learn More
Thermo Scientific™ Pre-Weigh Buffered Peptone Water (BPW) (ISO) Agar
Product Code. 15882477
Click to view available options
Quantity:
10 x Sachets for 8 L Medium
20 x Sachets for 1 L Medium
Unit Size:
1 set
Product Code. 15882477
Brand: Thermo Scientific™ PW10498L
Speed up your workflow when preparing medium for pre-enrichment of Salmonella species from foods with pre-weighed media sachets.
Speed up your workflow when preparing microbial culture medium for pre-enrichment of Salmonella species from food samples with Thermo Scientific™ Pre-weigh Buffered Peptone Water (ISO) Agar. The pre-weighed sachets of Thermo Scientific™ Oxoid™ dehydrated culture media are designed for single use when preparing 1 L or 8 L volumes of your medium. Save valuable time and start testing quickly by removing the requirement to weigh powder—just rip, tip and pour.
Pre-Weigh Buffered Peptone Water (ISO) sachets contain the familiar Thermo Scientific Oxoid Buffered Peptone Water (ISO) medium (Manf. Part No. CM1049) and are part of a range of our most frequently used and well-characterized dehydrated culture media, which is now available in handy pre-weighed foil packages.
Simply tear open the selected media sachet, add to the required volume of water, mix, and autoclave as usual.
  • Efficient: choose from a full range of pre-weighed media formats for reduced hands-on prep time and maximum productivity
  • Convenient: rip and tip sachets enable faster media production and simplified workflows
  • Compliant: proven and validated media formats deliver confidence and reliable performance
  • Productive: reduce routine dehydrated culture media handling and weighing to spend more time on value-added tasks.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Pre-Weigh Buffered Peptone Water (ISO)
Form Powder
Product Type Agar
Quantity 10 x Sachets for 8 L Medium
For Laboratory Use.