Description
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
- Broad chemical compatibility
- Biocompatible
- May be used with standard fingertight fittings
- Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
Specifications
Specifications
| For Use With | HPLC |
| Type | HPLC Tubing |
| Description | 0.030 in. ID; 5 ft. length; Green |
| For Use With (Application) | HPLC |
| Diameter (English) Inner | 0.030 in. |
| Diameter (Metric) Inner | 0.762 mm |
| Overall Length (English) | 5 ft. |
| Overall Length (Metric) | 1.524 m |
| Material | PEEK |