missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRDM16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15895813
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
15895813 100 μg 100µg
1 options
This item is not returnable. View return policy

Product Code. 15895813

Brand: Invitrogen™ 720206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with Monkey, Mouse, Dog.

PRDM16 is a zinc finger transcription factor and contains an N-terminal PR domain. The reciprocal translocation t(1;3)(p36;q21) occurs in a subset of myelodysplastic syndrome (MDS) and acute myeloid leukemia (AML). This gene is located near the 1p36.3 breakpoint and has been shown to be specifically expressed in the t(1:3)(p36, q21)-positive MDS/AML. The translocation results in the overexpression of a truncated version of this protein that lacks the PR domain, which may play an important role in the pathogenesis of MDS and AML. Recent studies have shown that PRDM16 normally acts as a Smad3 binding protein that may be important for the development of orofacial structures through modulation of the TGF-beta signaling pathway. Other experiments have indicated that PRDM16 controls a bidirectional cell fate switch between skeletal myoblasts and brown fat cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM16
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
Gene PRDM16
Gene Accession No. A2A935, Q9HAZ2
Gene Alias 5730557K01Rik; CMD1LL; csp1; Histone-lysine N-methyltransferase PRDM16; Kiaa1675; line 27; LVNC8; MDS1/EVI1-like gene 1; MEL1; PFM13; PR domain 16; PR domain containing 16; PR domain zinc finger protein 16; PR domain-containing protein 16; Prdm16; Transcription factor MEL1
Gene Symbols PRDM16
Host Species Rabbit
Immunogen Peptides corresponding to human PRDM16/MEL1 (236-251, 23-40).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100366024, 63976, 70673
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.