missing translation for 'onlineSavingsMsg'
Learn More

PRDM15 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25590-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894145

  • 3468.15 NOK / 150µL
Estimated Shipment: 14-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The exact function of PRDM15 remains unknown.
Specifications

Specifications

PRDM15
Polyclonal
Unconjugated
PRDM15
C21orf83, PFM15, PR domain containing 15, PRDM15, Zinc finger protein 298, ZNF298
Rabbit
Antigen Affinity Chromatography
RUO
63977
-20°C
Liquid
Western Blot, Immunoprecipitation
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P57071
PRDM15
PRDM15 Fusion Protein Ag22296
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers