Learn More
Pranlukast, MedChemExpress
Product Code. 16472086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472086 500 mg 500mg
16468216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16472086 Supplier Medchem Express Supplier No. HYB0290
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress Pranlukast is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively.
Pranlukast is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively.
Specifikationer
Chemical Name or Material Pranlukast
CAS 103177-37-3
Color White
Purity Grade Notes Research
Molecular Formula C27H23N5O4
Quantity 500 mg
Synonym ONO-1078
Solubility Information DMSO : 33.33 mg/mL (69.22 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=C2C(C(C=C(C3=NNN=N3)O2)=O)=CC=C1)C4=CC=C(OCCCCC5=CC=CC=C5)C=C4
Molecular Weight (g/mol) 481.5
Formula Weight 481.5
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Visa mer Visa mindre
Research purposes only
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.