Learn More
Pramoxine Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Pramoxine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Pramoxine Hydrochloride |
| CAS | 637-58-1 |
| Analyte or Component Names | Pramoxine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H27 N O3 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Morpholine, 4-[3-(4-butoxyphenoxy)propyl]-, hydrochloride (1:1)Morpholine, 4-[3-(4-butoxyphenoxy)propyl]-, hydrochloride (9CI), Morpholine, 4-[3-(p-butoxyphenoxy)propyl]-, hydrochloride (7CI,8CI), Pramoxine hydrochloride (6CI), 4-[3-(p-Butoxyphenoxy)propyl]morpholine hydrochloride, Pramacort, Pramocaine hydrochloride, Proctofoam, Tronolaine, Tronothan, Tronothane, Tronothane hydrochloride, 3389W95 |
| InChI Formula | InChI=1S/C17H27NO3.ClH/c1-2-3-12-20-16-5-7-17(8-6-16)21-13-4-9-18-10-14-19-15-11-18;/h5-8H,2-4,9-15H2,1H3;1H |
| SMILES | Cl.CCCCOc1ccc(OCCCN2CCOCC2)cc1 |
| IUPAC Name | 4-[3-(4-butoxyphenoxy)propyl]morpholine;hydrochloride |
| Show More |