Learn More
Pramipexole dihydrochloride, MedChemExpress
Product Code. p-200096828
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16447298
Brand: Medchem Express HY17355
MedChemExpress Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist, with Kis of 2.2 nM, 3.9 nM, 0.5 nM and 1.3 nM for D2-type receptor, D2, D3 and D4 receptors, respectively. Pramipexole dihydrochloride can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist, with Kis of 2.2 nM, 3.9 nM, 0.5 nM and 1.3 nM for D2-type receptor, D2, D3 and D4 receptors, respectively. Pramipexole dihydrochloride can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
Specifications
Chemical Name or Material Pramipexole dihydrochloride
Physical Form Solid
CAS 104632-25-9
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H19Cl2N3S
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information H2O : ≥ 50 mg/mL (175.90 mM)
SMILES NC(S1)=NC2=C1C[C@@H](NCCC)CC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 284.25
Formula Weight 284.25
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.