Learn More
Gestigkeit Harry Precision hotplate 200x280mm, type ET, without controller, 110°C
Brand:  Gestigkeit Harry PZ28-1ET
Precision hotplate 200x280mm, type ET, without controller, 110°C
Product Code. 10015030
  • ONLINE EXCLUSIVE PRICE
    12316.50 NOK
    10086.44 NOK / Pack of 1
    Save 2230.06 NOK
    18% Off
Estimated Shipment: 08-01-2027