missing translation for 'onlineSavingsMsg'
Learn More

PQR620, MedChemExpress

Product Code. p-200097302
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16424216 100 mg 100mg
16420499 1 mg 1mg
16415497 5 mg 5mg
16409456 50 mg 50mg
16402758 10 mg 10mg
5 options
This item is not returnable. View return policy

Product Code. 16424216

Brand: Medchem Express HY100026

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PQR620 is an orally bioavailable and selective brain penetrant inhibitor of mTORC1/2.

PQR620 is an orally bioavailable and selective brain penetrant inhibitor of mTORC1/2.

Specifications

Chemical Name or Material PQR620
CAS 1927857-56-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H25F2N7O2
Quantity 100 mg
Solubility Information DMSO : 6.4 mg/mL (14.37 mM; Need warming)
SMILES [H][C@]1(CC[C@]([H])2COC1)N2C3=NC(N4[C@]5([H])CC[C@]([H])4COC5)=NC(C6=CN=C(N)C=C6C(F)F)=N3
Molecular Weight (g/mol) 445.47
Formula Weight 445.47
Percent Purity 97.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.