Learn More
PQR620, MedChemExpress
Product Code. 16424216
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16424216 100 mg 100mg
16420499 1 mg 1mg
16415497 5 mg 5mg
16409456 50 mg 50mg
16402758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16424216 Supplier Medchem Express Supplier No. HY100026
MedChemExpress PQR620 is an orally bioavailable and selective brain penetrant inhibitor of mTORC1/2.
PQR620 is an orally bioavailable and selective brain penetrant inhibitor of mTORC1/2.
Specifications
Chemical Name or Material PQR620
CAS 1927857-56-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H25F2N7O2
Quantity 100 mg
Solubility Information DMSO : 6.4 mg/mL (14.37 mM; Need warming)
SMILES [H][C@]1(CC[C@]([H])2COC1)N2C3=NC(N4[C@]5([H])CC[C@]([H])4COC5)=NC(C6=CN=C(N)C=C6C(F)F)=N3
Molecular Weight (g/mol) 445.47
Formula Weight 445.47
Percent Purity 97.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease