Learn More
Bohlender™ BOLA™ PPS HT Screw Joint
Click to view available options
For Use With:
For 10mm tubing O.D.
For 12mm tubing O.D.
For 2mm tubing O.D.
For 3mm tubing O.D.
For 4mm tubing O.D.
For 6mm tubing O.D.
For 8mm tubing O.D.
Thread Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Description
- Black PPS screw cap
- Inner parts: PPS V-ring, FKM O-ring, PTFE tapered ring and sealing ring
- For tubing diameters under 3mm (not exposed to the medium)
- Good chemical resistance
- The medium is only exposed to PTFE
Specifications
Specifications
| Certifications/Compliance | FDA Conform |
| Material | PTFE |
| For Use With | For 8mm tubing O.D. |
| Min. Operating Temperature (Metric) | -50°C |
| Packaging Quantity | 1 Each |
| Thread Size | GL 25 |
| Color | Black |
| Maximum Pressure | 10 bar |
| Max. Operating Temperature (Metric) | 250°C |
| Product Type | HT Screw Joint |