Learn More
Abnova™ PPP6C (Human) Recombinant Protein
Human PPP6C full-length ORF ( AAH06990, 1 a.a. - 305 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005537-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAPLDLDKYVEIARLCKYLPENDLKRLCDYVCDLLLEESNVQPVSTPVTVCGDIHGQFYDLCELFRTGGQVPDTNYIFMGDFVDSGYYSLETFTYLLALKAKWPDRITLLRGNHESRQITQVYGFYDECQTKYGNANAWRYCTKVLDMLTVAALIDEQILCVHGGLSPDIKTLDQIRTIERNQEIPHKGAFCDLVWSDPEDVDTWAISPRGAGWLFGAKVTNEFVHINNLKLICRAHQLVHEGYKFMFDEKLVTVWSAPNYCYRCGNIASIMVFKDVNTREPKLFRAVPDSERVIPPRTTTPYFL
Specifications
AAH06990 | |
protein phosphatase 6, catalytic subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPP6C | |
GST |
5537 | |
Wheat germ expression system | |
10 μg | |
FLJ92648, MGC12249 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |