Learn More
PPP4C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10262-1-AP-20UL
 View more versions of this product
Product Code. 16856533
  • 1710.33 NOK / 20µL
Description
The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms.
Specifications
Specifications
PPP4C
Polyclonal
Unconjugated
PPP4C
PP X, PP4, PP4C, PPH3, PPP4, PPP4C, PPX, Protein phosphatase X
Rabbit
Antigen Affinity Chromatography
RUO
171366, 5531, 56420
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P60510, P97470, Q5BJ92
Ppp4c
PPP4C Fusion Protein Ag0356
20 μL
Primary
Rat, Mouse, Human