missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3CB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55148-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PPP3CB.
PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.Specifications
PPP3CB | |
Polyclonal | |
Unconjugated | |
PPP3CB | |
CALNA2, CALNB, CAM PRP catalytic subunit, CNA2, PPP3CB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19056, 24675, 5532 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16298, P20651, P48453 | |
Ppp3cb | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |