Learn More
PPM1D Mouse anti-Human, CoraLite 488, Clone: 2D6D5, Proteintech
Product Code. 17224746 Shop All Proteintech Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Artikelnummer. Quantity unitSize
17224746 100 μL 100µL
1 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17224746
missing translation for 'mfr': Proteintech CL48867440100UL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Mouse Monoclonal Antibody
The PPM1D antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human PPM1D. This antibody recognizes human antigen. The PPM1D antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Spezifikation
Antigen PPM1D
Applications Immunofluorescence
Classification Monoclonal
Clone 2D6D5
Concentration 1000 μg/mL
Conjugate CoraLite 488
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 8493
Target Species Human, Mouse, Rat
Form Liquid
Isotype IgG1
Mehr anzeigen Weniger anzeigen