missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPM1D (Human) Recombinant Protein
Human PPM1D partial ORF ( NP_003611, 496 a.a. - 605 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00008493-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: IGLVPTNSTNTVMDQKNLKMSTPGQMKAQEIERTPPTNFKRTLEESNSGPLMKKHRRNGLSRSSGAQPASLPTTSQRKNSVKLTMRRRLRGQKKIGNPLLHQHRKTVCVC
Specifications
NP_003611 | |
protein phosphatase 1D magnesium-dependent, delta isoform | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPM1D | |
GST |
8493 | |
Wheat germ expression system | |
10 μg | |
PP2C-DELTA, WIP1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |