missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPM1B (Human) Recombinant Protein
Human PPM1B full-length ORF ( AAH12002, 1 a.a. - 192 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005495-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSIVLVCFSNAPKVSDEAVKKDSELDKHLESRVEEIMEKSGEEGMPDLAHVMRILSAENIPNLPPGGGLAGKRNVIEAVYSRLNPHRESDGASDEAEESGSQGKLVEALRQMRINHRGNYRQLLEEMLTSYRLAKVEGEESPAEPAATATSSNSDAGNPVTMQESHTESESGLAELDSSNEDAGTKMSGEKI
Specifications
AAH12002 | |
protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPM1B | |
GST |
5495 | |
Wheat germ expression system | |
10 μg | |
MGC21657, PP2C-beta-X, PP2CB, PP2CBETA, PPC2BETAX | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |