missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15671-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cyclophilin family. Cyclophilins catalyze the cis-trans isomerization of peptidylprolyl imide bonds in oligopeptides. They have been proposed to act either as catalysts or as molecular chaperones in protein-folding events. Alternative splicing results in multiple transcript variants.Specifications
| PPIL3 | |
| Polyclonal | |
| Unconjugated | |
| PPIL3 | |
| Cyclophilin J, Cyclophilin like protein PPIL3, CYPJ, PPIase, PPIL3, Rotamase PPIL3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 301432, 53938, 70225 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q812D3, Q9D6L8, Q9H2H8 | |
| PPIL3 | |
| PPIL3 Fusion Protein Ag8137 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?