missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP2A alpha/beta Monoclonal Antibody (A0-G7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532920
This item is not returnable.
View return policy
Description
PP2A alpha/beta Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit.
Specifications
| PP2A alpha/beta | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P62714, P62715, P62716, P63330, P63331, P67775 | |
| PPP2CA, PPP2CB | |
| Synthetic peptide within Human PP2A(alpha+beta) aa 260-309. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot | |
| A0-G7 | |
| Unconjugated | |
| PPP2CB | |
| AI115466; D8Ertd766e; fb17h07; fc05c02; fk24a01; hypothetical protein LOC550585; PP2A; Pp2a1; Pp2a2; PP2A-alpha; PP2Abeta; PP2A-beta; PP2Ac; PP2CA; PP2Calpha; PP2CB; ppp2ca; ppp2caa; PPP2CB; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2 catalytic subunit alpha; protein phosphatase 2 catalytic subunit alpha a; protein phosphatase 2 catalytic subunit beta; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2,catalytic subunit alpha isozyme; protein phosphatase 2A alpha subunit; protein phosphatase 2A beta subunit; protein phosphatase 2A catalytic alpha subunit; protein phosphatase 2A catalytic subunit alpha isoform; protein phosphatase 2A catalytic subunit beta isoform; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase 2A, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-alpha; protein phosphatase-2A-beta; R75353; replication protein C; RP C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146; wu:fb17h07; wu:fc05c02; wu:fk24a01; zgc:110641; zgc:55808; zgc:77172 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 19052, 19053, 24672, 24673, 5515, 5516 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction