Learn More
Hirschmann™ Polypropylene Powder Funnels
Product Code. p-8011862
Change view
Click to view available options
Diameter (Metric)Top:
60mm
80mm
100mm
120mm
150mm
Unit Size:
Pack of 10
Pack of 5
Product Code. Diameter (Metric)Top unitSize
10680122 100mm Pack of 10
10601092 120mm Pack of 10
10620512 150mm Pack of 5
10194101 60mm Pack of 10
13158149 80mm Pack of 10
5 options
Product Code. 10680122
Brand: Hirschmann™ 7760103
Only null left
Add to basket
Add to basket
Ideal for use with powders and other large particles
  • Transparent
Specifications
Diameter (Metric)Top 100mm
Material Polypropylene
Height (Metric) 94 mm
Length (Metric) Stem 20mm
Quantity 10/Pk.