Learn More
Hirschmann™ Polypropylene Funnels
Product Code. p-8011861
Change view
Click to view available options
Diameter (Metric)Top:
40mm
75mm
100mm
120mm
150mm
Unit Size:
Pack of 12
Pack of 24
Product Code. Diameter (Metric)Top unitSize
10286581 120mm Pack of 12
10358231 100mm Pack of 12
10184341 150mm Pack of 12
10092751 40mm Pack of 24
10308671 75mm Pack of 12
5 options
Product Code. 10286581
Brand: Hirschmann™ 7750107
Only null left
Add to basket
Add to basket
For use with various laboratory applications
  • Transparent
Specifications
Diameter (Metric)Top 120mm
Material Polypropylene
Height (Metric) 180 mm
Length (Metric) Stem 90mm
Quantity 12/Pk.