Learn More
Multiroir™ Stackable Polypropylene Drawer
Product Code. 11707225
Click to view available options
Capacity (Metric):
25 L
Unit Size:
Each
Product Code. 11707225
Brand: Multiroir™ 18306
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Stack drawers atop each other for a dresser-like functional assembly that fits your storage and accessibility needs with the Multiroir™ Stackable Polypropylene Drawer.
  • White
  • Slide assembly;
  • Stackable
  • Lid included
Order Info
Color
Specifications
Capacity (Metric) 25 L
Color White
Material Polypropylene
Product Type Drawer
Quantity 1 Each
Dimensions (L x W x H) 410 x 320 x 220 mm
Length (Metric) 410 mm
Width (Metric) 320 mm
Height (Metric) 220 mm