Learn More
Bel Art™ Poxygrid™ Wire Test Tube Racks
Product Code. 30564744 Shop All SP Scienceware Products
Click to view available options
Color:
Blue
Green
Orange
Yellow
Positions:
48
60
72
108
150
Unit Size:
Each
Pack of 12
Pack of 24
Product Code. 30564744
Brand: Bel Art™ F187601150
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable
Specifications
Product Type Test Tube Rack
Array 10 x 15
Autoclavable Autoclavable
Color Blue
Holds Tubes (13 to 16 mm)
Material Epoxy-coated Steel
Positions 150
Shape Rectangular
Shelves 3 Tiers
Stackable Yes, When Empty
Sterility Non-sterile
Dimensions (L x W x H) 12 x 8.25 x 2.5 in. (30.5 x 21 x 6.4 cm)
Length (Metric) 305 mm
Length (English) 12 in.
Width (Metric) 209 mm
Width (English) 8.25 in.
Height (Metric) 64 mm
Height (English) 2.5 in.
Diameter (Metric) Cavity 150 mm
Diameter (Metric) Opening 16 mm