Learn More
Ansell™ PowerFlex™ 80-658 Series Aramid Fiber Heavy Weight Gloves
Product Code. 12657011 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. Size unitSize
12657011 6 12 pairs
11726723 10 12 pairs
12647011 11 12 pairs
11725834 8 12 pairs
11736723 9 12 pairs
15646906 7 12 pairs
6 options
Product Code. 12657011
Brand: Ansell™ 80658060
Only null left
Add to basket
Add to basket
Protects wearer with triple-layer construction comprised of steel, glass fiber and aramid fiber provides extreme resistance to cuts, splinters and burrs.
  • Mechanical protection gloves
  • Exceeds EN388 cut level 5.
  • Extra-long cuff offers extended safety for the forearm.
  • ¾ dipped and liquid repellent, shielding the skin from unpleasant liquid solutions.
  • Good heat contact resistance.
  • EN407 level-2 rating means safe handling of warm objects.
  • Natural rubber crinkle finish provides a firm grip.
  • Long life provides a highly cost-effective solution.
  • Heather green color helps mask dirt.
  • Excellent washability, with no decrease in comfort or fading.
Applications
  • Handling heavy loads, metal edges
  • Waste handling: recycling, sorting (glass, cans, metal pieces)
  • Handling glass sheets, sharp-edged objects
  • Handling metal sheets, cutting of dry, painted or galvanized pieces
TRUSTED_SUSTAINABILITY
Specifications
Back Material Dupont™ Kevlar™ aramid fibers
Certifications/Compliance CE Mark, Standard CATEGORY III EN 388 : X543, EN 407
Cuff Material Dupont™ Kevlar™ aramid fibers
Coating Material Natural Rubber
Color Blue
Cut Resistance Rating ANSI A5
Exterior Finish Crinkle
Hand Specific
Latex Free No
Material Cotton
Product Type Heavyweight Gloves
Size 6
Glove Cuff Style Long Knitwrist
Liner Material Natural Rubber Latex
Lining Lined
Palm Type 3/4 Dipped palm
Quantity 72 Pairs
Resistant To Good heat and cut resistance