Learn More
Ansell™ Powerflex™ 80-600
Product Code. 15891827 Shop All Ansell Products
Change view
Click to view available options
Size:
7
10
Unit Size:
12 pairs
Product Code. Size unitSize
15891827 7 12 pairs
15831837 10 12 pairs
2 options
Product Code. 15891827
Brand: Ansell™ 80600070
Only null left
Add to basket
Add to basket
Excellent cut resistance with comfort and a sure grip
  • Ansell's Powerflex™ 80-600 glove is the practical answer to the safe handling of glass, metal and plastics. It offers exceptional cut resistance and robust overall mechanical performance, plus superior comfort.
  • The Ansell Powerflex™ 80-600 glove is rated at the exceptionally high cut level of EN level 4. This high level of protection is achieved through the aramid fiber knitted liner and the natural rubber coating. This coating also stands up very well to abrasion, tearing and punctures.
  • The latex coating's raised crinkle finish gives the glove an excellent grip in both dry and wet conditions.
  • Silicone-free, the Powerflex™ 80-600 glove prevents fingerprints on metalised warm insulation glass, leaving surfaces clean and avoiding the need for extra cleaning work
TRUSTED_SUSTAINABILITY
Specifications
Back Material Aramid fiber, Natural rubber latex
Certifications/Compliance EN388:2016/3444C, EN407/X2XXXX
Classification Category III
Cleaning Procedure Washing temp 40° C
Coated Area Palm coated
Construction Knit
Cuff Material Aramid fiber, Natural rubber latex
Cut Style Newton
Description Natural rubber coated - open back and fingers with fully coated thumb -seamless dipped product on Dupont™ Kevlar™ aramid fiber liner
Coating Material Latex
Color Blue
Cut Resistance Rating 4
Disposable Reusable
Grade Industrial grade
Hand Specific
Latex Free No
Material Aramid Fiber, Natural Rubber, Latex
Powdered Powder Free
Product Type Mechanical/Cut Protection Gloves (Heavy-duty)
Size 7
Sterility Non-sterile
Style Powerflex 80-600
Thickness (Metric) 2.35 mm
Finger Style Full fingers
For Use With (Application) Mechanical protection
Gender Unisex
Glove Cuff Color Yellow
Glove Cuff Style Knit
Grip Design Rough
Liner Material Aramid fiber
Lining Knit Lining
Model 80-600
Packaging Type 12 pair in bag, 6 bags in a carton
Puncture Rating ANSI/ISEA 105-16 Level 4
Quantity 72 pair
Series Powerflex series
Tear Rating 4
Thumb Style Wing
Dimensions (L x W) 250 x 95 mm
Length (English) 250 mm
Width (Metric) 95 mm
Weight (Metric) 52 g