Learn More
Ansell™ Powerflex™ 80-400 Series Natural Rubber Latex Heavy Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
7
8
10
11
Unit Size:
12 pairs
Description
- Provides comfort and protection for the wearer in temperatures down to as low as -30°C, for short or intermittent contact.
- Terry loop acrylic liner that is in color
- Bright yellow liner profices extra high visibility in low or poor light conditions for additional safety
- Natural insulating latex coating
- Knitted construction
- Palm-coated finish
- Knitwrist cuff
Specifications
Specifications
| Back Material | Rubber Latex |
| Certifications/Compliance | Standard CATEGORY III EN 388 : X231, EN 407 : X2XXXX, EN 511 : 020, ANSI |
| Cuff Material | Natural Rubber Latex |
| Coating Material | Natural Rubber Latex |
| Color | Blue |
| Exterior Finish | No Texture |
| Hand | Specific |
| Material | Latex |
| Product Type | Heavyweight Gloves |
| Size | 10 |
| Show More |