Learn More
SI Analytics™ TZ 1858 Power Supply
Product Code. 10075230 Shop All SI Analytics Products
Change view
Click to view available options
Voltage:
100/220 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
10075230 100/220 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10075230 Supplier SI Analytics™ Supplier No. 285214970
  • ONLINE EXCLUSIVE PRICE
    1560.00 NOK
    1462.63 NOK / Pack of 1
    Save 97.37 NOK
    6% Off
 Restricted Item
Estimated Shipment: 27-11-2026
SI Analytics™ TZ 1858 Power Supply is for use with stirrer TM 128 and TM 135.
  • Power supply TZ 1858 100 to 220V
  • Output voltage 9V and 550 mA
  • For stirrer TM 128 and TM 135
TRUSTED_SUSTAINABILITY
Specifications
Product Type Power Supply
For Use With (Equipment) Stirrer TM 128, TM 135
Voltage 100/220 V
Amperage 550 mA