Learn More
Invitrogen™ Power Blotter Cassette XL
Description
Includes
Invitrogen Power Blotter XL Cassette
The Invitrogen™ Power Blotter Cassette is part of the Power Blotter System designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes.
This item is made available as a replacement or additional cassette for the Power Blotter Station.
The Power Blotter Cassette can be used with Power Blotter Select Transfer Stacks for rapid transfers, the Power Blotter 1-Step Transfer Buffer for semi-dry transfers, or alternatively with traditional buffers for longer semi-dry transfers.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| For Use With (Equipment) | PB0010 Power Blotter Station |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Mode of Transfer | Semi-Dry |
| Product Line | Power Blotter |
| Throughput | 2 Midi or 4 Mini Gels |
| Type | Blotter Cassette |
| Quantity | 1 cassette |
| Shipping Condition | Room Temperature |