Description
The Power Adapter for Multiskan Ease is the power supply for the Multiskan™ Ease Absorbance Microplate Reader, a modern easy-to-use absorbance reader designed to simplify both routine and advanced microplate-based assays. It enables consistent performance and safe operation in laboratory environments. Designed for global use, it features a universal input range of 100–240 VAC (50/60 Hz, 1.4 A) and enables a stable 24 V, 5 A output with a maximum power of 120 W.
Key Features
- Built-in protection against short circuit, overload, over-voltage, and over-temperature
- Fully enclosed, durable housing suitable for lab use
- Certified to IEC / EN / UL 62368-1 standards
- CE and UKCA marked
- FCC, ICES, and RoHS compliant
This power adapter is specifically compatible with the Thermo Scientific™ Multiskan™ Ease Absorbance Microplate Reader. Its robust construction supports long-term use in demanding laboratory settings, while comprehensive international safety and regulatory certifications help compliance and operational confidence across global markets.
Specifications
Specifications
| Product Type | External Power |
| For Use With (Equipment) | Multiskan Ease |
| Quantity | 1 Ea. |