Learn More
Buerkle™ Sampler PowderProof™
Product Code. 10693585 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
30 cm
60 cm
90 cm
120 cm
For Use With (Application):
Granules 2 to 4 mm dia.
Powders up to 2 mm dia.
Unit Size:
Each
Product Code. Length (Metric) For Use With (Application) unitSize
10693585 60 cm Powders up to 2 mm dia. Each
10041114 30 cm Powders up to 2 mm dia. Each
10706095 120 cm Granules 2 to 4 mm dia. Each
11877373 30 cm Granules 2 to 4 mm dia. Each
11887373 60 cm Granules 2 to 4 mm dia. Each
11897373 90 cm Granules 2 to 4 mm dia. Each
10346964 90 cm Powders up to 2 mm dia. Each
11807383 120 cm Powders up to 2 mm dia. Each
8 options
Product Code. 10693585
Brand: Buerkle™ 53020602
Only null left
Add to basket
Add to basket
Sampler PowderProof™ is the practically proven professional solution for mobile sampling of powders and granules.
  • Mobile sampling of powders and granules
  • Different lances and augers can be attached to the robust drive unit and swapped without any problems
  • For sampling powders and granules from drums, barrels, bags or sacks
  • PowderProof® with lance for horizontal use, such as sampling from sacks, with lance for vertical use, for sampling from open containers such as barrels, drums or bags
TRUSTED_SUSTAINABILITY
Specifications
Product Type PowderProof Sampler
Material Stainless Steel (V4A)
Includes 12 V Motor, two re-chargeable batteries, fast charger, clip for holding the bag and cleaning brush
For Use With (Application) Powders up to 2 mm dia.
Length (Metric) 60 cm