Learn More
Lenz Laborglasinstrumente™ Powder Flask with Conical Ground Joint
Product Code. 10204531
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10204531 250 mL Each
10090301 100 mL Each
11816074 2000 mL Each
11806104 1000 mL Each
11826204 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10204531 Supplier Lenz Laborglasinstrumente™ Supplier No. 03043049
  • ONLINE EXCLUSIVE PRICE
    1210.00 NOK
    962.66 NOK / Pack of 1
    Save 247.34 NOK
    20% Off
Estimated Shipment: 30-12-2026
Specifications
Material Duran Glass
Type Powder Flask
For Use With (Equipment) Rotary Evaporators
Volume (Metric) 250 mL
Diameter (Metric) 81 mm