Learn More
Asecos™ Powder-Coated Sheet Steel Base Cover
Product Code. 15295612 Shop All asecos Products
Change view
Click to view available options
Dimensions (D x W x H):
35 x 524 x 85 mm
35 x 818 x 85 mm
35 x 1118 x 85 mm
Unit Size:
Each
Product Code. Dimensions (D x W x H) unitSize
15295612 35 x 1118 x 85 mm Each
11959886 35 x 524 x 85 mm Each
15413643 35 x 818 x 85 mm Each
3 options
Product Code. 15295612
Brand: Asecos™ 30036
Interior equipment for use with safety storage cabinets.
  • Accessory item
  • Available in a range of sizes
  • Manufactured from powder-coated sheet steel
Specifications
Dimensions (D x W x H) 35 x 1118 x 85 mm
For Use With (Application) Provides protection
For Use With (Equipment) Q90 models
Surface Coating Powder-coated
Weight (Metric) 2 kg
Material Steel
Product Type Base Cover