Learn More
Lenz Laborglasintrumente Pouring Rings PBT Red
Description
- Material: PBT (red)
- Temperature resistance: up to 180 °C
Specifications
Specifications
| Color | Red |
| Material | PBT |
| Thread Size | GL 32 |