Learn More
Pour Point Standard IP15, ASTM D97, ISO 3016, BS 2000, Pt 15 (-30.5 °C) UKAS ISO 17034 Certified 250mL, Paragon Scientific™
Product Code. 15451850
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451850 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15451850 Supplier Paragon Scientific Supplier No. CRMUPPGO
  • ONLINE EXCLUSIVE PRICE
    3085.00 NOK
    2910.51 NOK / 250mL
    Save 174.49 NOK
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Pour Point Certified Reference Materials for the verification of laboratory instruments used in testing petroleum and derivative products for Pour Point to IP 15, ASTM D97, NFT 60-105.
  • Certified according to ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Certifications
Nominal values quoted
Specifications
UN Number UN 1202
Quantity 250 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • CRMU-PPGO
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
Nominal values quoted. Certified Values will vary from batch to batch.