Learn More
Pouds™ Wall Brackets
Description
Wall bracket for SHARPSGUARD™ mini, 1, 2.5, 3.75, 5 and com plus containers. Red.
Specifications
Specifications
| Color | Red |
| For Use With (Application) | Attaching sharps container to a wall |
| For Use With (Equipment) | Sharpsguard 7, 11.5, and 22 |