missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU5F2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | POU5F2 |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16844855
|
Proteintech
26362-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16834855
|
Proteintech
26362-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transcription factor that binds preferentially to the octamer motif (5′-ATGTTAAT-3′). May exert a regulatory function in meiotic events that are required for terminal differentiation of male germ cell.Specifications
| POU5F2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8N7G0 | |
| 134187 | |
| POU5F2 Fusion Protein Ag23803 | |
| Primary | |
| -20°C | |
| POU5F2 |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| POU5F2, SPRM 1, SPRM1 | |
| POU5F2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title