Learn More
POU2AF1 Mouse anti-Human, Clone: 3C5A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66659-1-IG-150UL
This item is not returnable.
View return policy
Description
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.Specifications
| POU2AF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16633 | |
| POU2AF1 | |
| POU2AF1 Fusion Protein Ag23613 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 3C5A7 | |
| Unconjugated | |
| POU2AF1 | |
| BOB 1, BOB1, OBF 1, OBF1, OCA B, OCAB, OCT binding factor 1, POU2AF1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5450 | |
| -20°C | |
| Liquid |