Learn More
Potassium Tetrachloroplatinum(II), TRC
Specifications
Specifications
| Chemical Name or Material | Potassium Tetrachloroplatinum(II) |
| CAS | 10025-99-7 |
| Molecular Formula | Cl4 Pt . 2 K |
| Quantity | 500 mg |
| Synonym | Platinate(2-)','tetrachloro-','potassium (1:2)','(SP-4-1)-, Platinate(2-)','tetrachloro-','dipotassium (8CI), Platinate(2-)','tetrachloro-','dipotassium','(SP-4-1)- (9CI), Potassium chloroplatinate(II) (6CI), Dipotassium platinum tetrachloride, Dipotassium tetrachloroplatinate, Dipotassium tetrachloroplatinate(2-), Dipotassium tetrachloroplatinate(II), NSC 175496, Platinous potassium chloride, Platinum potassium chloride (PtK2Cl4), Potassium chloroplatinate, Potassium chloroplatinate (K2PtCl4), Potassium platinochloride, Potassium platinous chloride, Potassium platinum(II) tetrachloride, Potassium tetrachloroplatinate, Potassium tetrachloroplatinate K2PtCl4, Potassium tetrachloroplatinate(2-), Potassium tetrachloroplatinate(II), Potassium tetrachloroplatinum(II), Tetrachloroplatinum(II) acid potassium salt |
| InChI Formula | InChI=1S/4ClH.2K.Pt/h4*1H;;;/q;;;;2*+1;+2/p-4 |
| SMILES | [K+].[K+].[Cl-][Pt+2]([Cl-])([Cl-])[Cl-] |
| Molecular Weight (g/mol) | 415.09 |
| Formula Weight | 412.7676 |
Product Title
Ser du en mulighed for forbedring?