Learn More
Potassium Tetrachloroplatinate(II), Extra Pure, SLR, Fisher Chemical
Product Code. 10274680 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Vial
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10274680 1 g Vial 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10274680 Supplier Fisher Chemical Supplier No. P/7170/43
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Melting Point 250°C
CAS 10025-99-7
Color Brown-Red
pH 4 to 5
Physical Form Solid
Quantity 1 g
Molecular Formula Cl4H4K2Pt
MDL Number MFCD00011378
Synonym potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
InChI Key JCAXKWJYKUEOFO-UHFFFAOYSA-N
SMILES Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]
IUPAC Name dipotassium tetrachloroplatinumdiuide
Molecular Weight (g/mol) 419.11
PubChem CID 61440
Formula Weight 415.1g/mol
Packaging Vial
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium tetrachloroplatinate(II)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .