missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrabromopalladate(II), Pd 20.0%

Product Code. p-7048893
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351968 1 g 1g
11361968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351968 Supplier Thermo Scientific Alfa Aesar Supplier No. 011883.03
Add to basket
This item is not returnable. View return policy

CAS: 13826-93-2 | Br4H4K2Pd | 508.26 g/mol

Potassium tetrabromopalladate(II) Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-93-2
Molecular Formula Br4H4K2Pd
Molecular Weight (g/mol) 508.26
MDL Number MFCD00049659
InChI Key PBPKCWAOLDCQKM-UHFFFAOYSA-N
Synonym potassium tetrabromopalladate ii, dipotassium tetrabromopalladium 2-, dipotassium tetrabromopalladate, dipotassium tetrabromopalladate 2-, palladate 2-, tetrabromo-, potassium 1:2, palladate 2-, tetrabromo-, dipotassium, sp-4-1, palladous potassium bromide, dipotassium tetrabromopalladiumdiuide, dipotassium ion tetrabromopalladiumdiuide
PubChem CID 61688
IUPAC Name dipotassium;tetrabromopalladium(2-)
SMILES [K+].[K+].Br.Br.Br.Br.[Pd--]

Specifications

Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 504.21
Concentration or Composition (by Analyte or Components) Pd 20.0%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Potassium tetrabromopalladate(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.