Learn More
Potassium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Product Code. p-7073218 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10169601
missing translation for 'mfr': Fisher Chemical J/8060/05
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Potassium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number 11409
Specifications
Color Colorless
pH 1
Physical Form Liquid
Quantity 100 mL
Packaging HDPE plastic bottle