Learn More
Potassium Permanganate, 0.002M, Chem Lab™
Product Code. 11993173
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 11993173
Brand: Chem-Lab CL05.1112.1000
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Specifications
Quantity 1 L
Concentration or Composition (by Analyte or Components) 0.002 mol/L
Density 1,00g/mL
Product Identifier
  • Potassium permanganate 0.002 mol/l
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Potassium permanganate