missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nitrate, TraceSELECT™, ≥99.995% (metals basis), Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
Unit Size:
100g
25g
Specifications
Specifications
| Melting Point | 335°C (1013hPa) |
| Color | Colorless |
| pH | 5.5 to 7.5 |
| Physical Form | Crystalline |
| Quantity | 25 g |
| UN Number | UN1486 |
| Formula Weight | 101.1g/mol |
| Percent Purity | ≥99.995% (metals basis) |
| Grade | TraceSELECT |
| Odor | Odorless |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction