Learn More
Potassium methanesulfonate, 99%
Product Code. 11358308
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358308 500 g 500g
11338308 25 g 25g
11348308 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11358308 Supplier Thermo Scientific Alfa Aesar Supplier No. 039505.36
Request Bulk or Custom Format
CAS: 2386-56-3 | CH3KO3S | 134.19 g/mol
Used in pharmaceutical, chemical and biological research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2386-56-3
Molecular Formula CH3KO3S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00070544
InChI Key XWIJIXWOZCRYEL-UHFFFAOYSA-M
Synonym potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1
PubChem CID 23666501
IUPAC Name potassium;methanesulfonate
SMILES CS(=O)(=O)[O-].[K+]
Specifications
Color White
Odor Odorless
Linear Formula KCH3SO3
Quantity 500 g
Beilstein 3914899
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 134.19
Percent Purity 99%
Physical Form Powder or Lumps
Chemical Name or Material Potassium methanesulfonate