Specifications
| Color Indicator | White to Blue/Black |
| Format | Reel |
| For Use With (Application) | Determination of Sulphit and Ssulphur dioxide |
| Quantity | 1 Each |
| Type | Qualitative Test Paper |
| Length (Metric) | 5000 mm |
| Width (Metric) | 7 mm |