missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate, 98%

Product Code. p-7023091
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Product Code. Quantity unitSize
11453217 100 g 100g
11463217 500 g 500g
11473217 2500 g 2500g
3 options
This item is not returnable. View return policy

Product Code. 11453217

Brand: Thermo Scientific Alfa Aesar A16162.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium iodate, 98%, IKO3, CAS Number-7758-05-6, iodic acid, potassium salt, kaliumjodat, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, potassium iodate, iodic acid hio3 , potassium salt, caswell no. 693a, potassium triodate, potassium iodine oxide kio3, unii-i139e44nhl | CAS: 7758-05-6 | IKO3 | 214.00 g/mol

Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

Solubility
Soluble in water, potassium iodide solution. Insoluble in alcohol, liquid ammonia and nitric acid.

Notes
Incompatible with aluminum, strong oxidizing agents, sulfides, peroxides, metals and reducing agents. Avoid heat, shock and friction.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Melting Point 560°C (decomposition)
Quantity 100 g
UN Number UN1479
Merck Index 14,7642
Solubility Information Soluble in water,potassium iodide solution. Insoluble in alcohol,liquid ammonia and nitric acid.
Formula Weight 214
Percent Purity 98%
Density 3.89
Chemical Name or Material Potassium iodate
compliance-icons compliance-icons
Product Identifier
  • Potassium iodate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Iodic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.