missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 45%, for HPLC, Honeywell Fluka™

Product Code. 15670680 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
4 x 25 mL
500 mL
Unit Size:
100mL
25mL
500mL
Pack of 4
Product Code. Quantity unitSize
15670680 4 x 25 mL Pack of 4
15698290 25 mL 25mL
15660680 100 mL 100mL
15680680 500 mL 500mL
4 options
This item is not returnable. View return policy

Product Code. 15670680

Brand: Honeywell-Fluka 035644X25ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for HPLC, ∼45%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
Concentration 45%
UN Number 1814
Density 1.450g/mL at 20°C
Quantity 4 x 25 mL
IUPAC Name potassium;hydroxide
Formula Weight 56.11g/mol
Chemical Name or Material Potassium Hydroxide Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.